Skip to Content
Merck
Cannabichromenic acid-D3 (CBCA-D3) solution 100 μg/mL (1% DIPEA and 0.05% Ascorbic Acid in Acetonitrile), certified reference material, ampule of 1 mL, Cerilliant®

Cannabichromenic acid-D3 (CBCA-D3) solution

Cannabichromenic acid-D3 (CBCA-D3) solution 100 μg/mL (1% DIPEA and 0.05% Ascorbic Acid in Acetonitrile), certified reference material, ampule of 1 mL, Cerilliant®
Empirical Formula (Hill Notation):
C22H27D3O4
Molecular Weight:
361.49

Products

Brand
Formula Weight
Manufacturer
Format
Quality Segment
Package Size
Reference Material Type
Special Grade