Skip to Content
Merck
Calcitonin human ≥97% (HPLC), powder

Calcitonin human

Calcitonin human ≥97% (HPLC), powder
Synonym(s):
Calcitonin M, human C carcinoma, Calcitonin, human reduced cyclic (1→7)-disulfide, Thyrocalcitonin, hCT
Empirical Formula (Hill Notation):
C151H226N40O45S3
CAS Number:
Molecular Weight:
3417.85
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Sterility
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing