![Bromhexine hydrochloride European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/217/975/94976680-5667-47f3-a393-add9106947d0/640/94976680-5667-47f3-a393-add9106947d0.png)
Bromhexine hydrochloride
![Bromhexine hydrochloride European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/217/975/94976680-5667-47f3-a393-add9106947d0/640/94976680-5667-47f3-a393-add9106947d0.png)
Synonym(s):
2-Amino-3,5-dibromo-N-cyclohexyl-N-methylbenzylamine hydrochloride, N-(2-Amino-3,5-dibromobenzyl)-N-methylcyclohexylamine hydrochloride, Bromohexine hydrochloride, Bromhexine hydrochloride
Empirical Formula (Hill Notation):
C14H20Br2N2 · HCl
CAS Number:
Molecular Weight:
412.59
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing