
Boc-Thr(Bzl)-OH

Synonym(s):
Boc-Thr(Bzl)-OH
Empirical Formula (Hill Notation):
C16H23NO5
CAS Number:
Molecular Weight:
309.36
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Functional Group
Application
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing