Skip to Content
Merck
Boc-6-Ahx-OH ≥99.0% (T)

Boc-6-Ahx-OH

Boc-6-Ahx-OH ≥99.0% (T)
Synonym(s):
6-(Boc-amino)caproic acid, 6-(Boc-amino)hexanoic acid, Boc-6-aminohexanoic acid
Linear Formula:
(CH3)3CO2CNH(CH2)5CO2H
CAS Number:
Molecular Weight:
231.29

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Markush Class
Markush Group
Amino Acid
Amino Acid Type
Reaction Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing