β-Mercaptoethanol, Molecular Biology Grade
Synonym(s):
2-Mercaptoethanol, β-Mercaptoethanol, 2-Hydroxyethylmercaptan, BME, Thioethylene glycol
Linear Formula:
HSCH2CH2OH
CAS Number:
Molecular Weight:
78.13
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing