Skip to Content
Merck
Bis(triphenylphosphine)dicarbonylnickel

Bis(triphenylphosphine)dicarbonylnickel

Bis(triphenylphosphine)dicarbonylnickel
Synonym(s):
Bis(triphenylphosphine)nickel(0)dicarbonyl, Dicarbonylbis(triphenylphosphine)nickel(0), Nickel(0)bis(triphenylphosphine) dicarbonyl
Linear Formula:
[(C6H5)3P]2Ni(CO)2
CAS Number:
Molecular Weight:
639.28
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing