Bisphenol F bis(2,3-dihydroxypropyl) ether
Synonym(s):
Bis[4-(2,3-dihydroxypropoxy)phenyl]methane
Empirical Formula (Hill Notation):
C19H24O6
CAS Number:
Molecular Weight:
348.39
Products
Formula Weight
Purity
Format
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing