Skip to Content
Merck
Biocytin ≥98% (TLC)

Biocytin

Biocytin ≥98% (TLC)
Synonym(s):
Bct, Nε-(+)-Biotinyl-L-lysine
Empirical Formula (Hill Notation):
C16H28N4O4S
CAS Number:
Molecular Weight:
372.48

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing