
Benoxathian hydrochloride

Synonym(s):
2-[[[2-(2,6-Dimethoxyphenoxy)ethyl]-amino]-methyl]-1,4-benzoxathian hydrochloride
Empirical Formula (Hill Notation):
C19H23NO4S · HCl
CAS Number:
Molecular Weight:
397.92
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing