Skip to Content
Merck
Benazepril Related Compound F Pharmaceutical Secondary Standard; Certified Reference Material

Benazepril Related Compound F

Benazepril Related Compound F Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
tert-Butyl (S)-3-amino-tetrahydro-2-oxo-1-benzazepine-1-acetate, (3S)-3-Amino-2,3,4,5-tetrahydro-2-oxo-1H-1-benzazepine-1-acetic acid tert-butyl ester
Empirical Formula (Hill Notation):
C16H22N2O3
CAS Number:
Molecular Weight:
290.36

Products

Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade