![Bathophenanthrolinedisulfonic acid disodium salt trihydrate for the spectrophotometric det. of Fe, ≥98.0% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/382/557/876f9db1-8cfc-48ad-ad65-7f903a1efebf/640/876f9db1-8cfc-48ad-ad65-7f903a1efebf.png)
Bathophenanthrolinedisulfonic acid disodium salt trihydrate
![Bathophenanthrolinedisulfonic acid disodium salt trihydrate for the spectrophotometric det. of Fe, ≥98.0% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/382/557/876f9db1-8cfc-48ad-ad65-7f903a1efebf/640/876f9db1-8cfc-48ad-ad65-7f903a1efebf.png)
Synonym(s):
4,7-Diphenyl-1,10-phenanthroline-disulfonic acid disodium salt trihydrate, Disodium bathophenanthrolinedisulfonate trihydrate
Empirical Formula (Hill Notation):
C24H14N2Na2O6S2 · 3H2O
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing