Astec® CHIROBIOTIC® R Chiral (5 μm) HPLC Columns
Products
Product Category
Brand
pH Value
Product Line
Separation Technique
ID (mm)
Length (cm)
Column Material
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing