Skip to Content
Merck
α-Pinene primary reference standard

α-Pinene

α-Pinene primary reference standard
Synonym(s):
(±)-2-Pinene, 2,6,6-Trimethylbicyclo[3.1.1]hept-2-ene
Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing