Skip to Content
CZ
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Anti-TRIM5 alpha antibody produced in rabbit
Products
Product Category
antibodies (3)
primary antibodies (3)
Brand
Sigma-Aldrich (3)
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
SAB3501194
Anti-TRIM5 alpha antibody produced in rabbit,
affinity isolated antibody
Expand
View Pricing
SAB3501193
Anti-TRIM5 alpha antibody produced in rabbit,
affinity isolated antibody
Expand
View Pricing
SAB3501195
Anti-TRIM5 alpha antibody produced in rabbit,
affinity isolated antibody
Expand
View Pricing