Anti-TF antibody produced in rabbit
Synonym(s):
Anti-β-1-Metal-binding globulin antibody produced in rabbit, Anti-Serotransferrin precursor antibody produced in rabbit, Anti-Siderophilin antibody produced in rabbit, Anti-Transferrin antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing