Anti-SMAD5 antibody produced in rabbit
Synonym(s):
Anti-Dwfc, Anti-JV5-1, Anti-MADH5, Anti-SMAD family member 5
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing