
Anti-NEUROD1 antibody produced in rabbit

Synonym(s):
Anti-NeuroD, Anti-NeuroD1, Anti-Neurogenic differentiation factor 1
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1