Skip to Content
Merck
Anti-HA−Peroxidase antibody, Mouse monoclonal clone HA-7, purified from hybridoma cell culture

Anti-HA−Peroxidase antibody, Mouse monoclonal

Anti-HA−Peroxidase antibody, Mouse monoclonal clone HA-7, purified from hybridoma cell culture
Synonym(s):
Monoclonal Anti-HA, Anti-HA, Anti-Influenza Hemagglutinin

Products

Product Category
Brand
Biological Source
Clonality
Physical Form
Antibody Form
Conjugate
Quality Segment
Package Size
Technique
Showing 1-1 of 1