![Anti-Digoxin–Alkaline Phosphatase antibody, Mouse monoclonal clone DI-22, purified from hybridoma cell culture](/deepweb/assets/sigmaaldrich/product/images/221/725/dc873758-3785-428f-ae83-861c6f12631f/640/dc873758-3785-428f-ae83-861c6f12631f.jpg)
Anti-Digoxin–Alkaline Phosphatase antibody, Mouse monoclonal
![Anti-Digoxin–Alkaline Phosphatase antibody, Mouse monoclonal clone DI-22, purified from hybridoma cell culture](/deepweb/assets/sigmaaldrich/product/images/221/725/dc873758-3785-428f-ae83-861c6f12631f/640/dc873758-3785-428f-ae83-861c6f12631f.jpg)
Synonym(s):
Monoclonal Anti-Digoxin
Products
Product Category
Brand
Biological Source
Clonality
Isotype
Physical Form
Recombinant Host
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing