Anti-CEP128 antibody produced in rabbit
Synonym(s):
Anti-C14orf145, Anti-Uncharacterized protein C14orf145 antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing