![Amprolium United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/161/635/4438885c-a4e2-4866-b585-fa73cd365dec/640/4438885c-a4e2-4866-b585-fa73cd365dec.png)
Amprolium
![Amprolium United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/161/635/4438885c-a4e2-4866-b585-fa73cd365dec/640/4438885c-a4e2-4866-b585-fa73cd365dec.png)
Synonym(s):
Amprolium hydrochloride, 1-([4-Amino-2-propyl-5-pyrimidinyl]methyl)-2-methylpyridinium chloride
Linear Formula:
C14H19N4Cl · HCl
CAS Number:
Molecular Weight:
315.24
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing