Skip to Content
Merck
Aminolevulinic Acid Related Compound B pharmaceutical secondary standard, certified reference material

Aminolevulinic acid Related Compound B

Aminolevulinic Acid Related Compound B pharmaceutical secondary standard, certified reference material
Synonym(s):
1,3-Dioxo-2-isoindolinelevulinic acid methyl ester, Methyl 5-(1,3-dioxoisoindolin-2-yl)-4-oxopentanoate
Empirical Formula (Hill Notation):
C14H13NO5
CAS Number:
Molecular Weight:
275.26

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade