![Amberlite™ MB-20 mixed bed ion exchange resin, regenerated](/deepweb/assets/sigmaaldrich/product/images/419/643/94eda8ed-e4e5-4333-8804-a6dd9f55b318/640/94eda8ed-e4e5-4333-8804-a6dd9f55b318.jpg)
Amberlite™ MB20 Ion Exchange Resin
![Amberlite™ MB-20 mixed bed ion exchange resin, regenerated](/deepweb/assets/sigmaaldrich/product/images/419/643/94eda8ed-e4e5-4333-8804-a6dd9f55b318/640/94eda8ed-e4e5-4333-8804-a6dd9f55b318.jpg)
Synonym(s):
AmberLite™ MB20 H/OH Mixed Bed Ion Exchange Resin hydrogen and hydroxide form
Products
Product Category
Brand
pH Value
Physical Form
Chemical Composition
Separation Technique
Quality Segment
Particle Size (µm)
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing