Skip to Content
CZ
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aluminum-magnesium-silicon alloy, Al98Mg1Si1
Products
Manufacturer
Physical Form
ID (mm)
Length (mm)
OD (mm)
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
GF48360225
Aluminum-magnesium-silicon alloy, Al98Mg1Si1,
200mm tube, 12.7mm outside diameter, 1.22mm wall thickness, 10.26mm inside diameter, hard
Expand
View Pricing
GF00386883
Aluminum-magnesium-silicon alloy, Al98Mg1Si1,
100mm tube, 12.7mm outside diameter, 1.22mm wall thickness, 10.26mm inside diameter, hard
Expand
View Pricing
GF40281623
Aluminum-magnesium-silicon alloy, Al98Mg1Si1,
500mm tube, 6.35mm outside diameter, 0.71mm wall thickness, 4.93mm inside diameter, hard
Expand
View Pricing
GF60104540
Aluminum-magnesium-silicon alloy, Al98Mg1Si1,
100mm tube, 25.4mm outside diameter, 3.25mm wall thickness, 18.9mm inside diameter, hard
Expand
View Pricing
GF13067726
Aluminum-magnesium-silicon alloy, Al98Mg1Si1,
200mm tube, 25.4mm outside diameter, 3.25mm wall thickness, 18.9mm inside diameter, hard
Expand
View Pricing
GF98229774
Aluminum-magnesium-silicon alloy, Al98Mg1Si1,
100mm tube, 6.35mm outside diameter, 0.71mm wall thickness, 4.93mm inside diameter, hard
Expand
View Pricing
GF83136911
Aluminum-magnesium-silicon alloy, Al98Mg1Si1,
1000mm tube, 25.4mm outside diameter, 3.25mm wall thickness, 18.9mm inside diameter, hard
Expand
View Pricing
GF33465090
Aluminum-magnesium-silicon alloy, Al98Mg1Si1,
500mm tube, 25.4mm outside diameter, 3.25mm wall thickness, 18.9mm inside diameter, hard
Expand
View Pricing
GF45027840
Aluminum-magnesium-silicon alloy, Al98Mg1Si1,
500mm tube, 12.7mm outside diameter, 1.22mm wall thickness, 10.26mm inside diameter, hard
Expand
View Pricing
GF35203092
Aluminum-magnesium-silicon alloy, Al98Mg1Si1,
1000mm tube, 6.35mm outside diameter, 0.71mm wall thickness, 4.93mm inside diameter, hard
Expand
View Pricing
GF28480833
Aluminum-magnesium-silicon alloy, Al98Mg1Si1,
1000mm tube, 12.7mm outside diameter, 1.22mm wall thickness, 10.26mm inside diameter, hard
Expand
View Pricing