Skip to Content
Merck
Aluminum cerium oxide nanopowder, ≤120 nm particle size (BET), 99% trace metals basis

Aluminum cerium oxide

Aluminum cerium oxide nanopowder, ≤120 nm particle size (BET), 99% trace metals basis
Synonym(s):
Cerium aluminate, Cerium aluminum oxide
Linear Formula:
AlCeO3
CAS Number:
Molecular Weight:
215.10

Products

Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1