![8-Bromoadenosine 3′,5′-cyclic monophosphate ≥97% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/417/568/6bd9d272-1705-41f0-a898-3545a3bff703/640/6bd9d272-1705-41f0-a898-3545a3bff703.png)
8-Bromoadenosine 3′,5′-cyclic monophosphate
![8-Bromoadenosine 3′,5′-cyclic monophosphate ≥97% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/417/568/6bd9d272-1705-41f0-a898-3545a3bff703/640/6bd9d272-1705-41f0-a898-3545a3bff703.png)
Synonym(s):
8-Br-A-3:5-MP, 8-Br-cAMP, 8-Bromo-cAMP
Empirical Formula (Hill Notation):
C10H11BrN5O6P
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing