![6-Hydroxyindole analytical standard](/deepweb/assets/sigmaaldrich/product/structures/235/634/c46f1d81-ff5c-4bf0-bac0-9ebefb586b9b/640/c46f1d81-ff5c-4bf0-bac0-9ebefb586b9b.png)
6-Hydroxyindole
![6-Hydroxyindole analytical standard](/deepweb/assets/sigmaaldrich/product/structures/235/634/c46f1d81-ff5c-4bf0-bac0-9ebefb586b9b/640/c46f1d81-ff5c-4bf0-bac0-9ebefb586b9b.png)
Synonym(s):
6-Indolol
Empirical Formula (Hill Notation):
C8H7NO
CAS Number:
Molecular Weight:
133.15
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing