6-Acetylmorphine-d3 solution
Synonym(s):
6-Acetylmorphine-N-methyl-d3 solution
Empirical Formula (Hill Notation):
C19D3H18NO4
CAS Number:
Molecular Weight:
330.39
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Research Area
Application
Package Size
Reference Material Type
Special Grade
Technique