![5-Phospho-D-ribose 1-diphosphate pentasodium salt ≥75% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/228/353/97efa769-3927-498c-b3cc-ba6fb8a51a6b/640/97efa769-3927-498c-b3cc-ba6fb8a51a6b.png)
5-Phospho-D-ribose 1-diphosphate pentasodium salt
![5-Phospho-D-ribose 1-diphosphate pentasodium salt ≥75% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/228/353/97efa769-3927-498c-b3cc-ba6fb8a51a6b/640/97efa769-3927-498c-b3cc-ba6fb8a51a6b.png)
Synonym(s):
α-D-Ribosyl diphosphate 5-phosphate pentasodium salt, 5-Phospho-α-D-ribosyl diphosphate, 5-Phosphoribosyl 1-pyrophosphate pentasodium salt, P-Rib-PP, PRPP pentasodium salt
Empirical Formula (Hill Notation):
C5H8Na5O14P3
CAS Number:
Molecular Weight:
499.98
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing