Skip to Content
Merck
[5,6]-Fullerene-C70 sublimed, ≥99% (HPLC)

[5,6]-Fullerene-C70

[5,6]-Fullerene-C70 sublimed, ≥99% (HPLC)
Synonym(s):
Buckminsterfullerene C70, Fullerene-C70
Empirical Formula (Hill Notation):
C70
CAS Number:
Molecular Weight:
840.75

Products

Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing