Skip to Content
Merck
4-Ethynylaniline 97%

4-Ethynylaniline

4-Ethynylaniline 97%
Synonym(s):
1-Amino-4-ethynylbenzene, P-APAC
Linear Formula:
HC≡CC6H4NH2
CAS Number:
Molecular Weight:
117.15

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing