Skip to Content
Merck
4,7-Diphenyl-1,10-phenanthroline

4,7-Diphenyl-1,10-phenanthroline

4,7-Diphenyl-1,10-phenanthroline
Synonym(s):
Bathophenanthroline, 4,7-Diphenyl-1,10-phenanthroline, BPhen
Empirical Formula (Hill Notation):
C24H16N2
CAS Number:
Molecular Weight:
332.40

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing