![3-Furancarboxaldehyde ≥97%](/deepweb/assets/sigmaaldrich/product/structures/175/837/4a65e2c1-dad3-4813-9415-353d2eff835f/640/4a65e2c1-dad3-4813-9415-353d2eff835f.png)
3-Furancarboxaldehyde
![3-Furancarboxaldehyde ≥97%](/deepweb/assets/sigmaaldrich/product/structures/175/837/4a65e2c1-dad3-4813-9415-353d2eff835f/640/4a65e2c1-dad3-4813-9415-353d2eff835f.png)
Synonym(s):
3-Furaldehyde
Empirical Formula (Hill Notation):
C5H4O2
CAS Number:
Molecular Weight:
96.08
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing