Skip to Content
Merck
3-(Bromomethyl)benzonitrile 95%

3-(Bromomethyl)benzonitrile

3-(Bromomethyl)benzonitrile 95%
Synonym(s):
α-Bromo-m-tolunitrile, 3-Cyanobenzyl bromide
Linear Formula:
BrCH2C6H4CN
CAS Number:
Molecular Weight:
196.04
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing