Skip to Content
Merck
3α,5β-Tetrahydroaldosterone ≥98% (CP)

3α,5β-Tetrahydroaldosterone

3α,5β-Tetrahydroaldosterone ≥98% (CP)
Synonym(s):
11β,18-Epoxy-3α,18,21-trihydroxy-5β-pregnan-20-one, 5β-Pregnan-11β,18-epoxy-3α,18,21-triol-20-one
Empirical Formula (Hill Notation):
C21H32O5
CAS Number:
Molecular Weight:
364.48

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing