3′,6-Disinapoylsucrose
Synonym(s):
DSS
Empirical Formula (Hill Notation):
C34H42O19
CAS Number:
Molecular Weight:
754.69
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing