Skip to Content
Merck
3,6-Dichloropyridazine 97%

3,6-Dichloropyridazine

3,6-Dichloropyridazine 97%
Synonym(s):
3,6-Dichloropyridazine
Empirical Formula (Hill Notation):
C4H2Cl2N2
CAS Number:
Molecular Weight:
148.98

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing