Skip to Content
Merck
2-Propanol 70 % (v/v) EMPROVE® EXPERT, USP

2-Propanol 70 % (v/v)

2-Propanol 70 % (v/v) EMPROVE® EXPERT, USP
Synonym(s):
Isopropanol, 2-Propanol solution, IPA, Isopropyl alcohol
CAS Number:

Products

Product Category
Brand
Product Line
Agency
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing