Skip to Content
Merck
2-Nonanol 99%

2-Nonanol

2-Nonanol 99%
Synonym(s):
Heptyl methyl carbinol, 2-Nonanol
Linear Formula:
CH3(CH2)6CH(OH)CH3
CAS Number:
Molecular Weight:
144.25

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Showing 1-3 of 3
Compare
Product Number
Description
Pricing