2-Nitrotoluene
Synonym(s):
1-Methyl-2-nitrobenzene, 2-Nitrotoluene
Linear Formula:
2-(NO2)C6H4CH3
CAS Number:
Molecular Weight:
137.14
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing