![2-Hydroxysaclofen ≥98% (TLC), solid](/deepweb/assets/sigmaaldrich/product/structures/175/108/42e4443d-e8e5-49b8-a832-6855f9f41eea/640/42e4443d-e8e5-49b8-a832-6855f9f41eea.png)
2-Hydroxysaclofen
![2-Hydroxysaclofen ≥98% (TLC), solid](/deepweb/assets/sigmaaldrich/product/structures/175/108/42e4443d-e8e5-49b8-a832-6855f9f41eea/640/42e4443d-e8e5-49b8-a832-6855f9f41eea.png)
Synonym(s):
3-Amino-2-(4-chlorophenyl)-2-hydroxypropanesulfonic acid
Empirical Formula (Hill Notation):
C9H12ClNO4S
CAS Number:
Molecular Weight:
265.71
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing