Skip to Content
Merck
2′-Deoxyguanosine 5′-triphosphate sodium salt hydrate ≥96% (HPLC)

2′-Deoxyguanosine 5′-triphosphate sodium salt hydrate

2′-Deoxyguanosine 5′-triphosphate sodium salt hydrate ≥96% (HPLC)
Synonym(s):
dGTP sodium salt hydrate
Empirical Formula (Hill Notation):
C10H16N5O13P3 · xNa+ · yH2O
CAS Number:
Molecular Weight:
507.18 (anhydrous free acid basis)

Products

Brand
Biological Source
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing