![2-Amino-1-(4-nitrophenyl)propane-1,3-diol British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/315/496/f9d070ce-6d9e-4dac-927d-0e201ea5f305/640/f9d070ce-6d9e-4dac-927d-0e201ea5f305.png)
2-Amino-1-(4-nitrophenyl)propane-1,3-diol
![2-Amino-1-(4-nitrophenyl)propane-1,3-diol British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/315/496/f9d070ce-6d9e-4dac-927d-0e201ea5f305/640/f9d070ce-6d9e-4dac-927d-0e201ea5f305.png)
Synonym(s):
(1S,2S)-(+)-2-Amino-1-(4-nitrophenyl)-1,3-propanediol, L-(+)-threo-2-Amino-1-(4-nitrophenyl)-1,3-propanediol
Linear Formula:
O2NC6H4CH(OH)CH(NH2)CH2OH
CAS Number:
Molecular Weight:
212.20
Products
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing