2-Acrylamido-2-methylpropanesulfonic acid
Synonym(s):
2-Acrylamido-2-methylpropanesulfonic acid
Empirical Formula (Hill Notation):
C7H13NO4S
CAS Number:
Molecular Weight:
207.25
Products
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing