Skip to Content
Merck
2,4,5-Trichlorophenol purum, ≥95.0% (GC)

2,4,5-Trichlorophenol

2,4,5-Trichlorophenol purum, ≥95.0% (GC)
Linear Formula:
Cl3C6H2OH
CAS Number:
Molecular Weight:
197.45
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing