Skip to Content
Merck
2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionic acid 95%

2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionic acid

2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionic acid 95%
Synonym(s):
Fenofibric acid
Empirical Formula (Hill Notation):
C17H15ClO4
CAS Number:
Molecular Weight:
318.75

Products

Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing