23-epi-26-Deoxyactein
Synonym(s):
26-Deoxyactein
Empirical Formula (Hill Notation):
C37H56O10
CAS Number:
Molecular Weight:
660.83
Products
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Package Size
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing