Skip to Content
Merck
1-Pyrenecarboxylic acid 97%

1-Pyrenecarboxylic acid

1-Pyrenecarboxylic acid 97%
Empirical Formula (Hill Notation):
C17H10O2
CAS Number:
Molecular Weight:
246.26

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing