Skip to Content
Merck
1-Acetylpiperidine-4-carboxylic acid 97%

1-Acetylpiperidine-4-carboxylic acid

1-Acetylpiperidine-4-carboxylic acid 97%
Synonym(s):
N-Acetylisonipecotic acid
Empirical Formula (Hill Notation):
C8H13NO3
CAS Number:
Molecular Weight:
171.19
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing